Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 2/CD49b Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB12331SP
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot.Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P17301 | |
| ITGA2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301 | |
| 25 μg | |
| Primary | |
| Detects human Integrin alpha 2/CD49b in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant mouse (rm) Integrin alpha 2 is observed and no cross-reactivity with recombinant human (rh) Integrin alpha 1, alpha 3, alpha 5, alpha 6, alpha 8, alpha 9, alpha 10, alpha 11, alpha L, alpha M, or alpha V is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 430907 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3673 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction