Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Integrin alpha 2/CD49b Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
$200.50 - $502.50
Specifications
| Antigen | Integrin alpha 2/CD49b |
|---|---|
| Clone | 430907 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12331SP
![]() |
R&D Systems
MAB12331SP |
25 μg |
Each for $200.50
|
|
|||||
MAB12331
![]() |
R&D Systems
MAB12331 |
100 μg |
Each for $502.50
|
|
|||||
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot.Specifications
| Integrin alpha 2/CD49b | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| P17301 | |
| 3673 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Integrin alpha 2/CD49b in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant mouse (rm) Integrin alpha 2 is observed and no cross-reactivity with recombinant human (rh) Integrin alpha 1, alpha 3, alpha 5, alpha 6, alpha 8, alpha 9, alpha 10, alpha 11, alpha L, alpha M, or alpha V is observed. |
| 430907 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| ITGA2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title