Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Integrin alpha 6 beta 1 Heterodimer Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7809100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7809100 100 μg
MAB7809SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7809100 Supplier R&D Systems Supplier No. MAB7809100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin alpha 6 beta 1 Monoclonal specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Integrin alpha 6 beta 1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2548B
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_000201
Gene Alias CD49f, ITGA6B, VLA-6
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.