Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Irisin/FNDC5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $503.00
Specifications
Antigen | Irisin/FNDC5 |
---|---|
Clone | 936410 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8880SP
![]() |
R&D Systems
MAB8880SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8880100
![]() |
R&D Systems
MAB8880100 |
100 μg |
Each for $503.00
|
|
|||||
Description
Irisin/FNDC5 Monoclonal specifically detects Irisin/FNDC5 in Human samples. It is validated for ELISA.Specifications
Irisin/FNDC5 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q8NAU1 | |
252995 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
936410 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2 | |
Chinese hamster ovary cell line CHO-derived recombinant human Irisin/FNDC5 Asp32-Glu143 Accession # Q8NAU1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Irisin/FNDC5 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title