Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ITIH2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB101991100
This item is not returnable.
View return policy
Description
ITIH2 Monoclonal antibody specifically detects ITIH2 in Human samples. It is validated for Western Blot, ELISASpecifications
ITIH2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
H2P, IGHEP2, Inter-Alpha (Globulin) Inhibitor H2, inter-alpha (globulin) inhibitor, H2 polypeptide, Inter-alpha-inhibitor heavy chain 2, Inter-alpha-trypsin inhibitor complex component II, inter-alpha-trypsin inhibitor heavy chain H2, Intin2, ITI heavy chain H2, ITI-H2, ITI-HC2, Serum-derived hyaluronan-associated protein, SHAP | |
Spodoptera frugiperda Sf 9 (baculovirus)-derived human ITIH2, Ser55-Asp702, Accession # P19823 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, ELISA | |
1023658 | |
Western Blot 2 ug/mL, ELISA | |
P19823 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3698 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction