Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ITIH2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $397.50

Specifications

Antigen ITIH2
Clone 1023658
Dilution Western Blot 2 ug/mL, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB101991SP
GREENER_CHOICE
View Documents
R&D Systems
MAB101991SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB10199110
GREENER_CHOICE
View Documents
R&D Systems
MAB101991100
100 μg
Each for $397.50
Only null left
Add to Cart
 
Description

Description

ITIH2 Monoclonal antibody specifically detects ITIH2 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

ITIH2
Western Blot 2 ug/mL, ELISA
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
H2P, IGHEP2, Inter-Alpha (Globulin) Inhibitor H2, inter-alpha (globulin) inhibitor, H2 polypeptide, Inter-alpha-inhibitor heavy chain 2, Inter-alpha-trypsin inhibitor complex component II, inter-alpha-trypsin inhibitor heavy chain H2, Intin2, ITI heavy chain H2, ITI-H2, ITI-HC2, Serum-derived hyaluronan-associated protein, SHAP
Spodoptera frugiperda Sf 9 (baculovirus)-derived human ITIH2, Ser55-Asp702, Accession # P19823
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1023658
Western Blot, ELISA
Unconjugated
Mouse
Human
P19823
3698
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.