Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KA1/GRIK4 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB113261100
This item is not returnable.
View return policy
Description
KA1/GRIK4/Glutamate Receptor KA1 Monoclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human samples. It is validated for ImmunohistochemistrySpecifications
KA1/GRIK4/Glutamate Receptor KA1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
EAA1, Excitatory amino acid receptor 1, GluK4, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, GRIK4, KA1GRIK | |
Human KA1/GRIK containing peptide, Accession # P42263 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
IgG |
Immunohistochemistry | |
2915C | |
Immunohistochemistry 5-15 ug/mL | |
P42263 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2900 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction