Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KA1/GRIK4 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Specifications
Antigen | KA1/GRIK4/Glutamate Receptor KA1 |
---|---|
Clone | 2915C |
Dilution | Immunohistochemistry 5-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
RD10000184
|
R&D Systems
MAB113261100 |
100 μg | N/A | N/A | N/A | ||||
MAB113261SP
|
R&D Systems
MAB113261SP |
25 μg | N/A | N/A | N/A | ||||
Description
KA1/GRIK4/Glutamate Receptor KA1 Monoclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human samples. It is validated for ImmunohistochemistrySpecifications
KA1/GRIK4/Glutamate Receptor KA1 | |
Immunohistochemistry 5-15 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
EAA1, Excitatory amino acid receptor 1, GluK4, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, GRIK4, KA1GRIK | |
Human KA1/GRIK containing peptide, Accession # P42263 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2915C | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P42263 | |
2900 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title