Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Kallikrein 12 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4034
This item is not returnable.
View return policy
Description
Kallikrein 12 Monoclonal specifically detects Kallikrein 12 in Human samples. It is validated for Western Blot.Specifications
| Kallikrein 12 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UKR0 | |
| KLK12 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0 | |
| 100 μg | |
| Primary | |
| Detects human Kallikrein 12 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Kallikrein 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 13, 14, 15, rhKallikrein beta 1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, rhThrombin, or rhuPA-1. | |
| Human | |
| Purified |
| Western Blot | |
| 364926 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686H1078, EC 3.4.21, EC 3.4.21.-, kallikrein 12, Kallikrein-like protein 5, kallikrein-related peptidase 12, KLK12, KLK-L5, KLKL5kallikrein-12, KLK-L5MGC42603 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 43849 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction