Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Kallikrein 12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7065903
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4034SP 25 μg
MAB4034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4034SP Supplier R&D Systems Supplier No. MAB4034SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kallikrein 12 Monoclonal specifically detects Kallikrein 12 in Human samples. It is validated for Western Blot.

Specifications

Antigen Kallikrein 12
Applications Western Blot
Classification Monoclonal
Clone 364926
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UKR0
Gene Alias DKFZp686H1078, EC 3.4.21, EC 3.4.21.-, kallikrein 12, Kallikrein-like protein 5, kallikrein-related peptidase 12, KLK12, KLK-L5, KLKL5kallikrein-12, KLK-L5MGC42603
Gene Symbols KLK12
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 Accession # Q9UKR0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43849
Test Specificity Detects human Kallikrein 12 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Kallikrein 1, 2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 13, 14, 15, rhKallikrein beta 1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, rhThrombin, or rhuPA-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.