Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KCC4/SLC12A7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067752
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9030SP 25 μg
MAB9030 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9030SP Supplier R&D Systems Supplier No. MAB9030SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KCC4/SLC12A7 Monoclonal specifically detects KCC4/SLC12A7 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen KCC4/SLC12A7
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 891526
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y666
Gene Alias DKFZp434F076, KCC4Electroneutral potassium-chloride cotransporter 4, K-Cl cotransporter 4, potassium/chloride transporter KCC4, SLC12A7, solute carrier family 12 (potassium/chloride transporters), member 7, solute carrier family 12 member 7
Gene Symbols SLC12A7
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC12A7 Met1-Ser1083 Accession # Q9Y666
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10723
Test Specificity Detects human SLC12A7 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.