Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Laminin alpha 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | Laminin alpha 2 |
---|---|
Clone | 1003531 |
Dilution | Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2143SP
![]() |
R&D Systems
MAB2143SP |
25 μg |
Each for $200.50
|
|
|||||
MAB2143100
![]() |
R&D Systems
MAB2143100 |
100 μg |
Each for $479.50
|
|
|||||
Description
Laminin alpha 2 Monoclonal specifically detects Laminin alpha 2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Laminin alpha 2 | |
Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
LAMA2, Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMM, LAMMLaminin-4 subunit alpha, Merosin, Merosin heavy chain | |
Chinese Hamster Ovary cell line, CHO-derived human Laminin alpha 2, Lys2145-Asn3122, Accession # P24043 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Laminin alpha 2 in direct ELISAs. |
1003531 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Human | |
P24043 | |
3908 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title