Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Laminin alpha 3/Laminin-5 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC21441G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC21441G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC21441G100 Supplier R&D Systems Supplier No. IC21441G100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Laminin alpha 3/Laminin-5 Monoclonal specifically detects Laminin alpha 3/Laminin-5 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Laminin alpha 3/Laminin-5
Applications Flow Cytometry
Classification Monoclonal
Clone 546215
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_000218
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Laminin alpha 3/Laminin-5, aa 21-1713, Accession # NP_000218
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human Laminin alpha 3/Laminin-5 in direct ELISAs and Western blots. In direct ELISAs, no cross reactivity with recombinant human (rh) Laminin alpha 1, beta 1, gamma 2, or recombinant mouse Laminin alpha 4 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.