Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human LECT2 Antibody, R&D Systems™
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Goat Polyclonal Antibody
$200.50 - $538.00
Specifications
| Antigen | LECT2 | 
|---|---|
| Concentration | LYOPH | 
| Dilution | Western Blot 0.1 ug/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
Description
LECT2 Polyclonal specifically detects LECT2 in Human samples. It is validated for Western Blot.Specifications
| LECT2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| chm2, chm-II, Chondromodulin II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628, MIM1 | |
| LECT2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O14960 | |
| 3950 | |
| E. coli-derived recombinant human LECT2 Gly19-Leu151 Accession # O14960 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human LECT2 in direct ELISAs and Western blots. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            