Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lefty-A Long and Short Isoforms Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7461
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7461 500 μg
MAB7461SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB7461 Supplier R&D Systems Supplier No. MAB7461
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lefty-A Monoclonal specifically detects Lefty-A in Human samples. It is validated for Western Blot.

Specifications

Antigen Lefty-A
Applications Western Blot
Classification Monoclonal
Clone 182525
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00292.2
Gene Alias EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, Lefta, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTY2, LeftyA, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A;transforming growth factor beta superfamily)
Gene Symbols LEFTY2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Lefty-A Phe78-Pro366 Accession # O00292.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7044
Test Specificity Detects both long and short forms of recombinant human (rh) Lefty-A in direct ELISAs and Western blots. No cross-reactivity with rhArtemin, recombinant mouse (rm) Artemin, recombinant drosophila DPP, rhGDNF, rhLAP, recombinant rat MIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, or rmLefty-1. The cross-reactivity of this antibody with rhLefty-B (96% amino acid sequence homology) was not determined.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.