Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Lefty-A Long and Short Isoforms Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $650.90
Specifications
| Antigen | Lefty-A |
|---|---|
| Clone | 182525 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Lefty-A Monoclonal specifically detects Lefty-A in Human samples. It is validated for Western Blot.Specifications
| Lefty-A | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, Lefta, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTY2, LeftyA, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A;transforming growth factor beta superfamily) | |
| LEFTY2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 182525 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00292.2 | |
| 7044 | |
| Mouse myeloma cell line NS0-derived recombinant human Lefty-A Phe78-Pro366 Accession # O00292.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects both long and short forms of recombinant human (rh) Lefty-A in direct ELISAs and Western blots. No cross-reactivity with rhArtemin, recombinant mouse (rm) Artemin, recombinant drosophila DPP, rhGDNF, rhLAP, recombinant rat MIS, rhNeurturin, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, or rmLefty-1. The cross-reactivity of this antibody with rhLefty-B (96% amino acid sequence homology) was not determined. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title