Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $686.23
Specifications
| Antigen | Legumain/Asparaginyl Endopeptidase |
|---|---|
| Clone | 312109 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB21992SP
![]() |
R&D Systems
MAB21992SP |
25 μg |
Each for $188.28
|
|
|||||
MAB21992
![]() |
R&D Systems
MAB21992 |
500 μg |
Each for $686.23
|
|
|||||
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| Legumain/Asparaginyl Endopeptidase | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34 | |
| LGMN | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 312109 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99538 | |
| 5641 | |
| NS0-derived recombinant human Legumain Val18-Tyr433 Accession # Q99538 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects both the pro and mature forms of human Legumain/Asparaginyl Endopeptidase in ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title