Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRA6/CD85b APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$245.00
Specifications
Antigen | ILT8/CD85b/LILRA6 |
---|---|
Clone | 921330 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB8656A025
![]() |
R&D Systems
FAB8656A025 |
25 Tests |
Each of 1 for $245.00
|
|
|||||
Description
ILT8/CD85b/LILRA6 Monoclonal specifically detects ILT8/CD85b/LILRA6 in Human samples. It is validated for Flow Cytometry.Specifications
ILT8/CD85b/LILRA6 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
79168 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human LILRA6/CD85b in direct ELISA. Detects HEK293 cells transfected with LILRA6/CD85b but not irrelevant transfectants. |
921330 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q6PI73 | |
LILRA6 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title