Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRB1/CD85j/ILT2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$162.80 - $432.50
Specifications
| Antigen | ILT2/CD85j/LILRB1 |
|---|---|
| Clone | 292305 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB20171P02
![]() |
R&D Systems
FAB20171P025 |
25 Tests |
Each for $162.80
|
|
|||||
FAB20171P10
![]() |
R&D Systems
FAB20171P100 |
100 Tests |
Each for $432.50
|
|
|||||
Description
ILT2/CD85j/LILRB1 Monoclonal specifically detects ILT2/CD85j/LILRB1 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT2/CD85j/LILRB1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7 | |
| LILRB1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ILT2/CD85j in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT4, rhILT5, rhLIR6 or rhLIR8 is observed. |
| 292305 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| Q8NHL6 | |
| 10859 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT2/CD85j Gly24-His458 Accession # Q8NHL6 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title