Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRB4/CD85k/ILT3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $538.00
Specifications
Antigen | ILT3/CD85k/LILRB4 |
---|---|
Clone | 293622 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
ILT3/CD85k/LILRB4 Monoclonal specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Western Blot.Specifications
ILT3/CD85k/LILRB4 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
ABM86208 | |
11006 | |
Mouse myeloma cell line NS0-derived recombinant human LILRB4/CD85k/ILT3 Pro17-His257 Accession # ABM86208 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human LILRB4/CD85k/ILT3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ILT2, 4 5, 6, rhLIR6, or rhLIR8 is observed. |
293622 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
LILRB4 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title