Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lipase I Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5909SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5909SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5909SP Supplier R&D Systems Supplier No. AF5909SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Lipase I Polyclonal specifically detects Lipase I in Human samples. It is validated for Western Blot.

Specifications

Antigen Lipase I
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6XZB0
Gene Alias Cancer/testis antigen 17, CT17, CT17membrane-associated phospholipase A1 beta, EC 3.1.1, EC 3.1.1.-, EC 3.1.1.3, EC 3.1.1.34, lipase, member I, LIPI, LPD lipase, LPDL, LPDLlipase member I, Membrane-associated phosphatidic acid-selective phospholipase A1-beta, mPA-PLA1 beta, PRED5
Gene Symbols LIPI
Host Species Goat
Immunogen E. coli-derived recombinant human Lipase I Arg14-His154 Accession # Q6XZB0
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149998
Test Specificity Detects human Lipase I in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.