Learn More
Human LRP-5 Intracellular Domain Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | LRP-5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 747619 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O75197 |
| Gene Alias | BMND1, BMND1OPTA1, EVR1, EVR4, HBM, low density lipoprotein receptor-related protein 5, low density lipoprotein receptor-related protein 7, low-density lipoprotein receptor-related protein 5, LR3VBCH2, LRP5, LRP-5, LRP7, LRP7exudative vitreoretinopathy 1, OPPG, OPS, OPTA1, osteoporosis pseudoglioma syndrome, VBCH2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.