Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LYVE-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB20893
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
LYVE-1 | |
Monoclonal | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Q9Y5Y7 | |
LYVE1 | |
Mouse myeloma cell line NS0-derived recombinant human LYVE-1 Ser24-Thr238 Accession # Q9Y5Y7 | |
500 μg | |
Primary | |
Detects human LYVE-1 in direct ELISAs. | |
Human | |
Purified |
Blockade of Receptor-ligand Interaction | |
537022 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10894 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction