Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LYVE-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | LYVE-1 |
---|---|
Clone | 537022 |
Concentration | LYOPH |
Dilution | Blockade of Receptor-ligand Interaction |
Applications | Blockade of Receptor-ligand Interaction |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB20893SP
![]() |
R&D Systems
MAB20893SP |
25 μg |
Each for $200.50
|
|
|||||
MAB20893
![]() |
R&D Systems
MAB20893 |
500 μg |
Each for $655.00
|
|
|||||
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
LYVE-1 | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Mouse | |
Human | |
Q9Y5Y7 | |
10894 | |
Mouse myeloma cell line NS0-derived recombinant human LYVE-1 Ser24-Thr238 Accession # Q9Y5Y7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human LYVE-1 in direct ELISAs. |
537022 | |
Blockade of Receptor-ligand Interaction | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
LYVE1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title