Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human M-CSF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB616SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB616SP 25 μg
MAB616100 100 μg
MAB616500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB616SP Supplier R&D Systems Supplier No. MAB616SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

M-CSF Monoclonal specifically detects M-CSF in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen M-CSF
Applications Western Blot, ELISA
Classification Monoclonal
Clone 21113
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930
Gene Symbols CSF1
Host Species Mouse
Immunogen E. coli-derived recombinant human M-CSF
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1435
Test Specificity Detects recombinant human (rh)M-CSF in Western blots. In sandwich ELISAs, less than 7% cross-reactivity was observed with rmM-CSF. No significant cross-reactivity or interference was observed with over 75 other cytokines tested.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.