Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human M-CSF R PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$210.00 - $444.00
Specifications
Antigen | M-CSFR/CD115 |
---|---|
Clone | 61708 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB329P025
![]() |
R&D Systems
FAB329P025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB329P100
![]() |
R&D Systems
FAB329P100 |
100 Tests |
Each for $444.00
|
|
|||||
Description
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for Flow Cytometry.Specifications
M-CSFR/CD115 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
CSF1R | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human M-CSF R in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) GM-CSF R alpha or rhGM-CSF R beta . |
61708 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
P07333.2 | |
1436 | |
Mouse myeloma cell line NS0-derived recombinant human M-CSF R Ile20-Glu512 (Pro54Ala) Accession # P07333.2 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title