Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human MBP DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human MBP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human MBP.
Specifications
Specifications
Quantity | 1 Kit (for 5 Plates) |
Species | Human |
Conjugate | Biotin |
Binding Target | MBP |
Assay Range | 234.40 to 15000 pg/mL |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 4155 |
Synonym | Hmbpr, MGC99675, mld, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein, shi |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction