Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MDGA2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5184
This item is not returnable.
View return policy
Description
MDGA2 Polyclonal specifically detects MDGA2 in Human samples. It is validated for Western Blot.Specifications
MDGA2 | |
Polyclonal | |
Unconjugated | |
Q7Z553 | |
MDGA2 | |
Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp931 Accession # Q7Z553 | |
100 μg | |
Primary | |
Detects human MDGA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human MDGA1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1 | |
Sheep | |
Affinity Purified | |
RUO | |
161357 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction