Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MDGA2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | MDGA2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
MDGA2 Polyclonal specifically detects MDGA2 in Human samples. It is validated for Western Blot.Specifications
MDGA2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1 | |
MDGA2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q7Z553 | |
161357 | |
Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp931 Accession # Q7Z553 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human MDGA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human MDGA1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title