Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MDGA2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5184SP
This item is not returnable.
View return policy
Description
MDGA2 Polyclonal specifically detects MDGA2 in Human samples. It is validated for Western Blot.Specifications
MDGA2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1 | |
Sheep | |
Affinity Purified | |
RUO | |
161357 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q7Z553 | |
MDGA2 | |
Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp931 Accession # Q7Z553 | |
25 μg | |
Primary | |
Detects human MDGA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human MDGA1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction