Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Meteorin-like/METRNL Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7161265
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB78672SP 25 μg
MAB78672100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB78672SP Supplier R&D Systems Supplier No. MAB78672SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Meteorin-like/METRNL
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 2441D
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 μ/mL, Immunocytochemistry 3-25 μ/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q641Q3
Gene Alias Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived human Meteorin-like/METRNL, Gln46-Asp311, Accession No. Q641Q3
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 284207
Test Specificity Detects human Meteorin-like/METRNL in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.