Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MFAP3L Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB11325SP
This item is not returnable.
View return policy
Description
MFAP3L Monoclonal antibody specifically detects MFAP3L in Human samples. It is validated for ImmunocytochemistrySpecifications
MFAP3L | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
HSD39, HSD-39, microfibrillar-associated protein 3-like, NYD-sp9, testis development protein NYD-SP9 | |
Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
Immunocytochemistry | |
1059107 | |
Immunocytochemistry 8-25 ug/mL | |
O75121 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9848 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction