Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MFAP3L Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$177.10 - $454.00
Specifications
Antigen | MFAP3L |
---|---|
Clone | 1059107 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11325100
![]() |
R&D Systems
MAB11325100 |
100 μg |
Each for $454.00
|
|
|||||
MAB11325SP
![]() |
R&D Systems
MAB11325SP |
25 μg |
Each for $177.10
|
|
|||||
Description
MFAP3L Monoclonal antibody specifically detects MFAP3L in Human samples. It is validated for ImmunocytochemistrySpecifications
MFAP3L | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
HSD39, HSD-39, microfibrillar-associated protein 3-like, NYD-sp9, testis development protein NYD-SP9 | |
Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1059107 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
O75121 | |
9848 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title