Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICA Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | MICA |
---|---|
Clone | 159233 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB13002SP
![]() |
R&D Systems
MAB13002SP |
25 μg |
Each for $200.50
|
|
|||||
MAB13002100
![]() |
R&D Systems
MAB13002100 |
100 μg |
Each for $549.50
|
|
|||||
MAB13002500
![]() |
R&D Systems
MAB13002500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
MICA Monoclonal specifically detects MICA in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
MICA | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q29983 | |
100507436 | |
BaF3 mouse pro-B cell line transfected with human MICA Accession # Q29983 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human MICA in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MICB is observed. |
159233 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ60820, MGC111087, PERB11.1 | |
MICA | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title