Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICL/CLEC12A Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2946
This item is not returnable.
View return policy
Description
MICL/CLEC12A Polyclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
MICL/CLEC12A | |
Polyclonal | |
Unconjugated | |
NP_612210 | |
CLEC12A | |
Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A isoform a Thr67-Ala265 Accession # NP_612210 | |
100 μg | |
Primary | |
Detects human MICL/CLEC12A in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse MICL is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
Goat | |
Affinity Purified | |
RUO | |
160364 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction