Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MICL/CLEC12A Antibody, R&D Systems™

Goat Polyclonal Antibody
Specifications
Antigen | MICL/CLEC12A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
MICL/CLEC12A Polyclonal specifically detects MICL/CLEC12A in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
MICL/CLEC12A | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
CLEC12A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
NP_612210 | |
160364 | |
Mouse myeloma cell line NS0-derived recombinant human MICL/CLEC12A isoform a Thr67-Ala265 Accession # NP_612210 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human MICL/CLEC12A in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant mouse MICL is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title