Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MMP-1 Antibody, R&D Systems™
GREENER_CHOICE

Recombinant Monoclonal Antibody

$241.00 - $561.50

Specifications

Antigen MMP-1
Clone 36665R
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Knockout Validated
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB901RSP
GREENER_CHOICE
View Documents
R&D Systems
MAB901RSP
25 μg
Each for $241.00
Only null left
Add to Cart
 
MAB901R100
GREENER_CHOICE
View Documents
R&D Systems
MAB901R100
100 μg
Each for $561.50
Only null left
Add to Cart
 
Description

Description

MMP-1 Monoclonal specifically detects MMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications

Specifications

MMP-1
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Knockout Validated
Unconjugated
Mouse
Human
CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1
MMP1
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
36665R
Monoclonal
Purified
RUO
P03956
4312
IgG1
Protein A or G purified from cell culture supernatant
Detects pro and active forms of human MMP-1.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.