Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB9022
This item is not returnable.
View return policy
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MMP-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
4313 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunoprecipitation | |
101724 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P08253 | |
MMP2 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-2 Ala30-Cys660 Accession # P08253 | |
500 μg | |
Primary | |
Detects the pro and active forms of human MMP-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MMP-1, -3, -7, -8, -9, -10, -12, or -13 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction