Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $655.00
Specifications
Antigen | MMP-2 |
---|---|
Clone | 101724 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
MMP-2 Monoclonal specifically detects MMP-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MMP-2 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase) | |
MMP2 | |
IgG2b | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
101724 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P08253 | |
4313 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-2 Ala30-Cys660 Accession # P08253 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects the pro and active forms of human MMP-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MMP-1, -3, -7, -8, -9, -10, -12, or -13 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title