Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Adenosine Deaminase/ADA Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Catalog No. AF7048T
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7048T 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF7048T Supplier R&D Systems Supplier No. AF7048T
Only null left
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Adenosine Deaminase/ADA Polyclonal antibody specifically detects Adenosine Deaminase/ADA in Human,Mouse samples. It is validated for Western Blot,Gene Knock-Out

Specifications

Antigen Adenosine Deaminase/ADA
Applications Western Blot, Gene Knock-Out
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation Supplied 0.2mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias ADA, ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Adenosine Deaminase/ADA, Met1-Leu363, Accession # P00813
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.