Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Bovine Integrin alpha 6/CD49f Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA13501T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA13501T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA13501T100 Supplier R&D Systems Supplier No. FAB13501T100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 6/CD49f
Applications Flow Cytometry
Classification Monoclonal
Clone GoH3
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6, ITGA6B, Platelet gpl, VLA-6
Gene Symbols ITGA6
Host Species Rat
Immunogen Mouse mammary tumor cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3655
Test Specificity Detects human, mouse, and bovine Integrin alpha 6/CD49f. Recognizes an epitope in the extracellular domain of the Integrin alpha 6 subunit.
Target Species Human, Mouse, Bovine
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.