Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CTLA-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | CTLA-4 |
---|---|
Clone | 1003705 |
Dilution | Western Blot 2 μg/mL, Agonist Activity 0.05-0.4 μg/mL |
Applications | Western Blot, In vitro Assay |
Classification | Monoclonal |
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human, Mouse samples. It is validated for Western Blot, Agonist Activity.Specifications
CTLA-4 | |
Western Blot 2 μg/mL, Agonist Activity 0.05-0.4 μg/mL | |
Monoclonal | |
Supernatant | |
Human, Mouse | |
P16410 | |
1493 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1003705 | |
Western Blot, In vitro Assay | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Chinese Hamster Ovary cell line, CHO-derived human CTLA-4, Ala37-Phe162, Accession # P16410 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CTLA-4 in direct ELISAs. Detects human and moue CTLA-4 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title