Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CXCL12/SDF-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 38 publications
$214.00 - $2070.00
Specifications
Antigen | CXCL12/SDF-1 |
---|---|
Clone | 79014 |
Dilution | Neutralization |
Applications | Neutralization |
Classification | Monoclonal |
Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Neutralization.Specifications
CXCL12/SDF-1 | |
Neutralization | |
Monoclonal | |
Mouse | |
Human, Mouse | |
Q6ICW0 | |
6387 | |
E. coli-derived recombinant human CXCL12/SDF-1 alpha Lys22-Lys89 Accession # Q6ICW0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse CXCL12/SDF-1 alpha and human CXCL12/SDF-1 beta . |
79014 | |
Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSF, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF1, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
CXCL12 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title