Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Doublecortin/DCX Antibody, R&D Systems™

Sheep Antibody
$200.50 - $573.00
Specifications
| Antigen | Doublecortin |
|---|---|
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF10025SP
![]() |
R&D Systems
AF10025SP |
25 μg |
Each for $200.50
|
|
|||||
AF10025100
![]() |
R&D Systems
AF10025100 |
100 μg |
Each for $573.00
|
|
|||||
Description
Doublecortin specifically detects Doublecortin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Doublecortin | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Sheep | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DBCN, Doublecortex, doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, LISX, X-linked (doublecortin) | |
| E.coli-derived recombinant human Doublecortin/DCX, Met116-Thr293, Accession # O43602 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Doublecortin/DCX in direct ELISAs and Western blots. Detects mouse Doublecortin/DCX in immunohistochemistry. |
| Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL | |
| Polyclonal | |
| Lyophilized | |
| Human, Mouse | |
| O43602 | |
| 1641 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title