Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Frizzled-2 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 1 publication

$214.00 - $2350.00

Specifications

Antigen Frizzled-2
Clone 202102
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Immunoprecipitation
Applications Western Blot, Immunoprecipitation
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1307SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1307SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB1307050
GREENER_CHOICE
View Documents
R&D Systems
MAB1307050
50 μg
Each for $292.00
Only null left
Add to Cart
 
MAB1307500
GREENER_CHOICE
View Documents
R&D Systems
MAB1307500
500 μg
Each for $2,350.00
Only null left
Add to Cart
 
Description

Description

Frizzled-2 Monoclonal specifically detects Frizzled-2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Frizzled-2
LYOPH
Western Blot, Immunoprecipitation
Unconjugated
Rat
Human, Mouse
Q9JIP6
2535
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Frizzled-1, -3, -4, -6, -7, -8, or rhFrizzled-5.
202102
Western Blot 1 ug/mL, Immunoprecipitation
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2
FZD2
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.