Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse HMGB1/HMG-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Clone | 1002147 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB16902SP
![]() |
R&D Systems
MAB16902SP |
25 μg |
Each for $200.50
|
|
|||||
MAB16902100
![]() |
R&D Systems
MAB16902100 |
100 μg |
Each for $491.00
|
|
|||||
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Immunocytochemistry.Specifications
HMGB1/HMG-1 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human, Mouse | |
P09429 | |
3146 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1002147 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
Synthetic peptide containing human HMGB1, Thr85-Gly166, Accession # P09429 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human HMGB1 in direct ELISAs. Detects human and mouse HMGB1 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title