Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-5 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 13 publications
$200.50 - $2110.00
Specifications
Antigen | IL-5 |
---|---|
Clone | TRFK5 |
Dilution | Neutralization 0.004-0.015 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-reported, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Neutralization, Flow Cytometry, CyTOF, ELISA |
Classification | Monoclonal |
Description
IL-5 Monoclonal specifically detects IL-5 in Human, Mouse samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-reported, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair).Specifications
IL-5 | |
Neutralization 0.004-0.015 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-reported, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Rat | |
Human, Mouse | |
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
IL5 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
TRFK5 | |
Neutralization, Flow Cytometry, CyTOF, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
3567 | |
Mouse T cell clone-derived partially purified recombinant mouse IL-5 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse IL-5 in ELISAs. In sandwich immunoassays with BAM6051 as the detection antibody, no cross-reactivity with recombinant human (rh) IL-3, rhGM-CSF, or recombinant mouse (rm) IL-5 is observed. In sandwich ELISAs with BAM705 as the detection antibody, no cross-reactivity with rhIL-5, rmIL-3, or rmGM-CSF is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title