Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Nurr1/NGFI-B beta/NR4A2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
$227.50 - $596.50
Specifications
Antigen | Nurr1/NGFI-B beta/NR4A2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Polyclonal |
Description
Nurr1/NGFI-B beta/NR4A2 Polyclonal specifically detects Nurr1/NGFI-B beta/NR4A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Nurr1/NGFI-B beta/NR4A2 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse | |
human homolog of, NGFI-B beta, NOT, NR4A2, nuclear receptor subfamily 4, group A, member 2, orphan nuclear receptor NR4A2, Orphan nuclear receptor NURR1, T-cell nuclear receptor NOT, TINUR, transcriptionally inducible nuclear receptor related 1, Transcriptionally-inducible nuclear receptor | |
NR4A2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
RUO | |
Q06219 | |
4929 | |
E. coli-derived recombinant mouse Nurr1/NGFI-B beta/NR4A2 Val332-Lys558 Accession # Q06219 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human and mouse Nurr1/NGFI-B beta/NR4A2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title