Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PKA RII alpha Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $491.00
Specifications
| Antigen | Protein Kinase A Regulatory Subunit II alpha |
|---|---|
| Clone | 2394D |
| Dilution | Western Blot 0.5 μg/mL, Immunohistochemistry 0.3-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8000SP
![]() |
R&D Systems
MAB8000SP |
25 μg |
Each for $214.00
|
|
|||||
MAB8000100
![]() |
R&D Systems
MAB8000100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Protein Kinase A Regulatory Subunit II alpha Monoclonal specifically detects Protein Kinase A Regulatory Subunit II alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Protein Kinase A Regulatory Subunit II alpha | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 0.3-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse | |
| P13861 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2394D | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Synthetic peptide containing Human/Mouse PKA RII alpha , Accession # P13861 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PKA RII alpha in direct ELISAs. Detects human and mouse PKA RII alpha in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title