Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat alpha-Actinin 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
| Antigen | alpha-Actinin 1 |
|---|---|
| Clone | 910301 |
| Concentration | LYOPH |
| Dilution | Western Blot 0.2 ug/mL, Simple Western 2 ug/mL |
| Applications | Western Blot |
Description
alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
| alpha-Actinin 1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P12814 | |
| 87 | |
| E. coli-derived recombinant human alpha-Actinin 1 Asn272-Thr500 Accession # P12814 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human alpha-Actinin 1 in ELISA, and human, mouse and rat alpha-Actinin 1 in Western Blot. |
| 910301 | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| actinin 1 smooth muscle, actinin, alpha 1, ACTN1, alphaActinin 1, alpha-Actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, BDPLT15, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
| ACTN1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title